Assay Details
Gene Symbol: PPP2R2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261139.1 | NP_001248068.1 | 697 | 77 | ||
XM_015137939.2 | XP_014993425.1 | 657 | 77 | |||
XM_015137940.2 | XP_014993426.1 | 722 | 77 | |||
XM_015137941.2 | XP_014993427.1 | 824 | 77 | |||
GenBank mRNA | JU319783.1 | - | 697 | 77 | ||
JU319784.1 | - | 697 | 77 | |||
JU319785.1 | - | 697 | 77 | |||
JV621099.1 | - | 927 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
