Assay Details
| Gene Symbol: LOC698860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845698.1 | XP_028701531.1 | 2081 | 79 | ||
| XM_028845699.1 | XP_028701532.1 | 1921 | 79 | |||
| XM_028845700.1 | XP_028701533.1 | 738 | 79 | |||
| GenBank mRNA | JU334459.1 | - | 357 | 79 | ||
| JU475548.1 | - | 273 | 79 | |||
| JV043323.1 | - | 443 | 79 | |||
More Information
Set Membership: |
|

LOC698860
Genomic Map