Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123640.2 | XP_014979126.2 | 1158 | 75 | ||
XM_015123641.2 | XP_014979127.2 | 1041 | 75 | |||
XM_015123643.2 | XP_014979129.2 | 819 | 75 | |||
XM_028839056.1 | XP_028694889.1 | 497 | 75 | |||
XM_028839057.1 | XP_028694890.1 | 702 | 75 | |||
XM_028839058.1 | XP_028694891.1 | 596 | 75 | |||
XM_028839059.1 | XP_028694892.1 | 424 | 75 | |||
XM_028839060.1 | XP_028694893.1 | 405 | 75 | |||
XM_028839061.1 | XP_028694894.1 | 996 | 75 | |||
GenBank mRNA | JU322954.1 | - | 285 | 75 | ||
JV544581.1 | - | 285 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028839061.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
