Assay Details
| Gene Symbol: RANBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260864.2 | NP_001247793.1 | 440 | 62 | ||
| XM_015149727.1 | XP_015005213.1 | 532 | 62 | |||
| XM_028827452.1 | XP_028683285.1 | 590 | 62 | |||
| GenBank mRNA | JU335751.1 | - | 432 | 62 | ||
| JU476636.1 | - | 448 | 62 | |||
| JV047819.1 | - | 441 | 62 | |||
| JV527776.1 | - | 424 | 62 | |||
| JV624606.1 | - | 460 | 62 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RANBP1
Genomic Map