Assay Details
Gene Symbol: LCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134101.2 | XP_014989587.1 | 1352 | 108 | ||
XM_015134108.2 | XP_014989594.1 | 1398 | 108 | |||
XM_015134119.2 | XP_014989605.1 | 1174 | 108 | |||
XM_015134128.2 | XP_014989614.1 | 1213 | 108 | |||
GenBank mRNA | JU475562.1 | - | 1178 | 108 | ||
JU486467.1 | - | 134 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
