Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086708.4 | XP_001086708.3 | 584 | 62 | ||
| XM_015121150.2 | XP_014976636.1 | 587 | 62 | |||
| XM_015121309.2 | XP_014976795.1 | 537 | 62 | |||
| XM_015121362.2 | XP_014976848.1 | 534 | 62 | |||
| GenBank mRNA | JU484232.1 | - | 258 | 62 | ||
| JV565046.1 | - | 399 | 62 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SZRD1
Genomic Map