Assay Details
Gene Symbol: ARCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265905.1 | NP_001252834.1 | 578 | 98 | ||
GenBank mRNA | JU322140.1 | - | 597 | 98 | ||
JU475503.1 | - | 597 | 98 | |||
JV044373.1 | - | 578 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
