Assay Details
Gene Symbol: YAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002799773.2 | XP_002799819.2 | 631 | 81 | ||
XM_002799774.3 | XP_002799820.2 | 319 | 81 | |||
XM_002799775.3 | XP_002799821.2 | 319 | 81 | |||
XM_015115542.2 | XP_014971028.1 | 613 | 81 | |||
XM_015115543.2 | XP_014971029.1 | 319 | 81 | |||
XM_015115544.2 | XP_014971030.1 | 613 | 81 | |||
XM_015115545.2 | XP_014971031.1 | 613 | 81 | |||
XM_015115546.2 | XP_014971032.1 | 319 | 81 | |||
XM_015115548.2 | XP_014971034.1 | 613 | 81 | |||
XM_015115549.2 | XP_014971035.1 | 319 | 81 | |||
XM_015115550.2 | XP_014971036.1 | 319 | 81 | |||
XM_015115551.2 | XP_014971037.1 | 319 | 81 | |||
GenBank mRNA | JV603436.1 | - | 423 | 81 | ||
JV635909.1 | - | 338 | 81 | |||
JV635910.1 | - | 338 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
