Assay Details
| Gene Symbol: DCUN1D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260996.1 | NP_001247925.1 | 528 | 92 | ||
| XM_015115529.2 | XP_014971015.1 | 662 | 92 | |||
| GenBank mRNA | JU320939.1 | - | 561 | 92 | ||
| JU477483.1 | - | 529 | 92 | |||
| JV048432.1 | - | 528 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015115529.2 transcripts.
This assay is in the 5'UTR region of the XM_015115529.2 transcripts.
Set Membership: |
|

DCUN1D5
Genomic Map