Assay Details
Gene Symbol: SEPTIN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113104.2 | XP_014968590.1 | 851 | 149 | ||
XM_015113105.2 | XP_014968591.1 | 620 | 149 | |||
XM_015113107.2 | XP_014968593.1 | 965 | 149 | |||
XM_015113109.2 | XP_014968595.1 | 629 | 149 | |||
XM_015113110.2 | XP_014968596.1 | 401 | 149 | |||
XM_015113111.2 | XP_014968597.1 | 776 | 149 | |||
XM_015113113.2 | XP_014968599.1 | 805 | 149 | |||
XM_015113114.2 | XP_014968600.1 | 736 | 149 | |||
XM_028832290.1 | XP_028688123.1 | 486 | 149 | |||
XM_028832292.1 | XP_028688125.1 | 619 | 149 | |||
XM_028832293.1 | XP_028688126.1 | 851 | 149 | |||
XM_028832294.1 | XP_028688127.1 | 776 | 149 | |||
GenBank mRNA | JU321498.1 | - | 555 | 149 | ||
JU470655.1 | - | 364 | 149 | |||
JV045546.1 | - | 449 | 149 | |||
JV638942.1 | - | 458 | 149 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
