Assay Details
Gene Symbol: SLC35B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257835.1 | NP_001244764.1 | 615 | 90 | ||
XM_015135456.2 | XP_014990942.1 | 743 | 90 | |||
XM_015135457.2 | XP_014990943.1 | 833 | 90 | |||
XM_015135458.2 | XP_014990944.1 | 1107 | 90 | |||
XM_015135459.2 | XP_014990945.1 | 780 | 90 | |||
XM_015135461.2 | XP_014990947.1 | 773 | 90 | |||
XM_028846664.1 | XP_028702497.1 | 743 | 90 | |||
XM_028846665.1 | XP_028702498.1 | 1082 | 90 | |||
GenBank mRNA | JU334756.1 | - | 615 | 90 | ||
JU471317.1 | - | 982 | 90 | |||
JV044521.1 | - | 764 | 90 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
