Assay Details
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193713.2 | NP_001180642.2 | 1591 | 131 | ||
GenBank mRNA | JU325921.1 | - | 987 | 131 | ||
JV066185.1 | - | 1601 | 131 | |||
JV600236.1 | - | 1524 | 131 | |||
JV656441.1 | - | 1514 | 131 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
