Assay Details
| Gene Symbol: LSM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141326.2 | XP_014996812.2 | 495 | 74 | ||
| XM_028849864.1 | XP_028705697.1 | 492 | 74 | |||
| GenBank mRNA | JU480483.1 | - | 421 | 74 | ||
More Information
Set Membership: |
|

LSM11
Genomic Map