Assay Details
Gene Symbol: SLCO5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099565.3 | XP_001099565.2 | 1737 | 62 | ||
XM_015145540.2 | XP_015001026.1 | 1737 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
