Assay Details
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105066.4 | XP_001105066.1 | 832 | 62 | ||
GenBank mRNA | JU326572.1 | - | 430 | 62 | ||
JV565053.1 | - | 462 | 62 | |||
JV603400.1 | - | 517 | 62 | |||
JV670477.1 | - | 521 | 62 | |||
KT751174.1 | - | 434 | 62 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
