Assay Details
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096861.4 | XP_001096861.1 | 293 | 68 | ||
GenBank mRNA | JU323328.1 | - | 229 | 68 | ||
JV047925.1 | - | 225 | 68 | |||
JV636319.1 | - | 225 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
