Assay Details
| Gene Symbol: SPAG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118895.2 | XP_014974381.2 | 3282 | 93 | ||
| XM_028836138.1 | XP_028691971.1 | 3947 | 93 | |||
| GenBank mRNA | JV492320.1 | - | 3148 | 93 | ||
| JV670844.1 | - | 1338 | 93 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPAG5
Genomic Map