Assay Details
| Gene Symbol: CCND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150849.2 | XP_015006335.1 | 1032 | 57 | ||
| GenBank mRNA | JU321222.1 | - | 1001 | 57 | ||
| JU477698.1 | - | 971 | 57 | |||
| JV043469.1 | - | 950 | 57 | |||
| JV043470.1 | - | 950 | 57 | |||
| JV726200.1 | - | 1007 | 57 | |||
| JV726201.1 | - | 1007 | 57 | |||
More Information
Set Membership: |
|

CCND2
Genomic Map