Assay Details
| Gene Symbol: MTCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194094.1 | NP_001181023.1 | 441 | 75 | ||
| XM_015128571.1 | XP_014984057.1 | 326 | 75 | |||
| GenBank mRNA | JU472130.1 | - | 341 | 75 | ||
| JV652057.1 | - | 276 | 75 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MTCP1
Genomic Map