Assay Details
| Gene Symbol: RAB30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261123.1 | NP_001248052.1 | 107 | 63 | ||
| XM_015115282.2 | XP_014970768.1 | 331 | 63 | |||
| XM_015115284.2 | XP_014970770.1 | 720 | 63 | |||
| XM_015115287.2 | XP_014970773.1 | 558 | 63 | |||
| XM_015115289.2 | XP_014970775.1 | 372 | 63 | |||
| XM_015115290.2 | XP_014970776.1 | 496 | 63 | |||
| XM_028832593.1 | XP_028688426.1 | 994 | 63 | |||
| XM_028832594.1 | XP_028688427.1 | 814 | 63 | |||
| XM_028832595.1 | XP_028688428.1 | 814 | 63 | |||
| GenBank mRNA | JU334030.1 | - | 205 | 63 | ||
| JV046779.1 | - | 107 | 63 | |||
| JV648511.1 | - | 114 | 63 | |||
More Information
Set Membership: |
|

RAB30
Genomic Map