Assay Details
| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266452.1 | NP_001253381.1 | 364 | 137 | ||
| XM_028848039.1 | XP_028703872.1 | 285 | 137 | |||
| GenBank mRNA | JU334046.1 | - | 398 | 137 | ||
| JU475215.1 | - | 364 | 137 | |||
| JV712313.1 | - | 412 | 137 | |||
More Information
Set Membership: |
|

GLRB
Genomic Map