Assay Details
Gene Symbol: IRAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136101.1 | NP_001129573.1 | 825 | 100 | ||
XM_015151459.2 | XP_015006945.2 | 1365 | 100 | |||
XM_015151461.2 | XP_015006947.2 | 1001 | 100 | |||
XM_015151462.2 | XP_015006948.1 | 896 | 100 | |||
XM_015151463.2 | XP_015006949.1 | 853 | 100 | |||
XM_028828838.1 | XP_028684671.1 | 1959 | 100 | |||
XM_028828839.1 | XP_028684672.1 | 1311 | 100 | |||
XM_028828840.1 | XP_028684673.1 | 2026 | 100 | |||
XM_028828841.1 | XP_028684674.1 | 2200 | 100 | |||
XM_028828842.1 | XP_028684675.1 | 1813 | 100 | |||
GenBank mRNA | EU204922.1 | - | 825 | 100 | ||
JU334803.1 | - | 903 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
