Assay Details
Gene Symbol: MYLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113601.4 | XP_001113601.1 | 2105 | 69 | ||
XM_015126075.2 | XP_014981561.1 | 1656 | 69 | |||
XM_015126076.2 | XP_014981562.1 | 1575 | 69 | |||
XM_015126077.2 | XP_014981563.1 | 1227 | 69 | |||
XM_015126078.2 | XP_014981564.1 | 1150 | 69 | |||
XM_028840637.1 | XP_028696470.1 | 1528 | 69 | |||
XM_028840638.1 | XP_028696471.1 | 1109 | 69 | |||
XM_028840639.1 | XP_028696472.1 | 1145 | 69 | |||
XM_028840640.1 | XP_028696473.1 | 1247 | 69 | |||
GenBank mRNA | JV665407.1 | - | 452 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
