Assay Details
| Gene Symbol: DLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092830.4 | XP_001092830.2 | 4741 | 65 | ||
| XM_015144785.2 | XP_015000271.1 | 3227 | 65 | |||
| XM_015144786.2 | XP_015000272.1 | 3474 | 65 | |||
| XM_028852365.1 | XP_028708198.1 | 4613 | 65 | |||
| GenBank mRNA | JU332491.1 | - | 4485 | 65 | ||
| JV565052.1 | - | 3007 | 65 | |||
More Information
Set Membership: |
|

DLC1
Genomic Map