Assay Details
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193843.3 | NP_001180772.2 | 874 | 96 | ||
XM_001089505.3 | XP_001089505.2 | 1248 | 96 | |||
XM_015124961.2 | XP_014980447.1 | 722 | 96 | |||
XM_028838394.1 | XP_028694227.1 | 738 | 96 | |||
GenBank mRNA | JU334209.1 | - | 654 | 96 | ||
JV649529.1 | - | 961 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
