Assay Details
Gene Symbol: TMEM164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261008.1 | NP_001247937.1 | 603 | 67 | ||
XM_015127983.2 | XP_014983469.1 | 796 | 67 | |||
XM_015127985.2 | XP_014983471.1 | 1833 | 67 | |||
XM_015127986.2 | XP_014983472.1 | 181 | 67 | |||
XM_028841783.1 | XP_028697616.1 | 825 | 67 | |||
XM_028841784.1 | XP_028697617.1 | 825 | 67 | |||
XM_028841785.1 | XP_028697618.1 | 1066 | 67 | |||
XM_028841786.1 | XP_028697619.1 | 826 | 67 | |||
XM_028841787.1 | XP_028697620.1 | 1100 | 67 | |||
XM_028841788.1 | XP_028697621.1 | 1874 | 67 | |||
XM_028841789.1 | XP_028697622.1 | 511 | 67 | |||
XM_028841790.1 | XP_028697623.1 | 526 | 67 | |||
GenBank mRNA | JU472732.1 | - | 603 | 67 | ||
JV635543.1 | - | 764 | 67 | |||
JV637407.1 | - | 182 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015127986.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
