Assay Details
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101030.4 | XP_001101030.2 | 607 | 91 | ||
GenBank mRNA | JU324371.1 | - | 477 | 91 | ||
JV645146.1 | - | 501 | 91 | |||
JV709423.1 | - | 498 | 91 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
