Assay Details
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266720.2 | NP_001253649.1 | 444 | 135 | ||
XM_015147634.2 | XP_015003120.1 | 496 | 135 | |||
XM_015147636.2 | XP_015003122.1 | 795 | 135 | |||
XM_028853355.1 | XP_028709188.1 | 405 | 135 | |||
XM_028853356.1 | XP_028709189.1 | 496 | 135 | |||
GenBank mRNA | JU322797.1 | - | 548 | 135 | ||
JU472827.1 | - | 537 | 135 | |||
JV045627.1 | - | 545 | 135 | |||
JV565064.1 | - | 444 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
