Assay Details
| Gene Symbol: VAMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261784.1 | NP_001248713.1 | 495 | 104 | ||
| XM_028841769.1 | XP_028697602.1 | 693 | 104 | |||
| XM_028841770.1 | XP_028697603.1 | 690 | 104 | |||
| XM_028841771.1 | XP_028697604.1 | 642 | 104 | |||
| GenBank mRNA | JU320414.1 | - | 618 | 104 | ||
| JU320415.1 | - | 495 | 104 | |||
| JU478345.1 | - | 625 | 104 | |||
| JV704568.1 | - | 497 | 104 | |||
More Information
Set Membership: |
|

VAMP7
Genomic Map