Assay Details
| Gene Symbol: RNF217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107488.4 | XP_001107488.1 | 1301 | 66 | ||
| GenBank mRNA | JU331222.1 | - | 398 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF217| Gene Symbol: RNF217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107488.4 | XP_001107488.1 | 1301 | 66 | ||
| GenBank mRNA | JU331222.1 | - | 398 | 66 | ||
Set Membership: |
|