Assay Details
| Gene Symbol: ZNRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194453.2 | NP_001181382.1 | 1048 | 65 | ||
| XM_015126610.2 | XP_014982096.1 | 1253 | 65 | |||
| XM_028840546.1 | XP_028696379.1 | 1253 | 65 | |||
| GenBank mRNA | JU321754.1 | - | 784 | 65 | ||
| JU471267.1 | - | 1013 | 65 | |||
| JV045170.1 | - | 1245 | 65 | |||
More Information
Set Membership: |
|

ZNRF1
Genomic Map