Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193848.3 | NP_001180777.3 | 383 | 85 | ||
| XM_028852152.1 | XP_028707985.1 | 275 | 85 | |||
| GenBank mRNA | JU484113.1 | - | 302 | 85 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VIRMA
Genomic Map