Assay Details
Gene Symbol: B4GALT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257845.1 | NP_001244774.1 | 492 | 68 | ||
XM_015113116.2 | XP_014968602.1 | 555 | 68 | |||
XM_028846222.1 | XP_028702055.1 | 654 | 68 | |||
GenBank mRNA | JU336895.1 | - | 492 | 68 | ||
JU472293.1 | - | 479 | 68 | |||
JV044881.1 | - | 509 | 68 | |||
JV566268.1 | - | 497 | 68 | |||
JV664391.1 | - | 516 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
