Assay Details
| Gene Symbol: KIAA0513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194581.2 | NP_001181510.1 | 1026 | 81 | ||
| XM_015126714.2 | XP_014982200.1 | 1084 | 81 | |||
| GenBank mRNA | JU332256.1 | - | 978 | 81 | ||
| JU478014.1 | - | 807 | 81 | |||
| JV600224.1 | - | 994 | 81 | |||
| JV667752.1 | - | 995 | 81 | |||

KIAA0513
Genomic Map