Assay Details
| Gene Symbol: PPP1R1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266974.1 | NP_001253903.1 | 994 | 60 | ||
| XM_015119510.2 | XP_014974996.1 | 1607 | 60 | |||
| XM_015119511.1 | XP_014974997.1 | 688 | 60 | |||
| XM_028835450.1 | XP_028691283.1 | 501 | 60 | |||
| GenBank mRNA | JU336668.1 | - | 875 | 60 | ||
| JU471256.1 | - | 994 | 60 | |||
| JV724915.1 | - | 1002 | 60 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP1R1B
Genomic Map