Assay Details
| Gene Symbol: CDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261470.1 | NP_001248399.1 | 239 | 109 | ||
| XM_015147555.2 | XP_015003041.2 | 347 | 109 | |||
| GenBank mRNA | JU472131.1 | - | 239 | 109 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDK1
Genomic Map