Assay Details
| Gene Symbol: TRIM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266149.1 | NP_001253078.1 | 142 | 77 | ||
| GenBank mRNA | JV565940.1 | - | 135 | 77 | ||
| JV629373.1 | - | 91 | 77 | |||
| JV667481.1 | - | 142 | 77 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRIM46
Genomic Map