Assay Details
Gene Symbol: POLDIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261713.2 | NP_001248642.1 | 854 | 70 | ||
XM_015118842.2 | XP_014974328.1 | 892 | 70 | |||
GenBank mRNA | JU333919.1 | - | 821 | 70 | ||
JU475106.1 | - | 825 | 70 | |||
JV046712.1 | - | 854 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
