Assay Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193708.2 | NP_001180637.1 | 834 | 104 | ||
| GenBank mRNA | JU321543.1 | - | 775 | 104 | ||
| JU473530.1 | - | 762 | 104 | |||
| JV044197.1 | - | 763 | 104 | |||
| JV565952.1 | - | 763 | 104 | |||
More Information
Set Membership: |
|

HSPA13
Genomic Map