Assay Details
| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261683.1 | NP_001248612.1 | 1780 | 71 | ||
| XM_015145735.2 | XP_015001221.2 | 1952 | 71 | |||
| GenBank mRNA | JU321202.1 | - | 1780 | 71 | ||
| JU327315.1 | - | 1575 | 71 | |||
| JU471353.1 | - | 1782 | 71 | |||
| JV043566.1 | - | 1790 | 71 | |||
More Information
Set Membership: |
|

LRP12
Genomic Map