Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087466.4 | XP_001087466.4 | 2685 | 90 | ||
XM_015139364.2 | XP_014994850.2 | 2723 | 90 | |||
XM_028848823.1 | XP_028704656.1 | 2797 | 90 | |||
XM_028848824.1 | XP_028704657.1 | 2682 | 90 | |||
GenBank mRNA | JU483201.1 | - | 2656 | 90 | ||
JV710477.1 | - | 2576 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
