Assay Details
| Gene Symbol: UQCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194866.3 | NP_001181795.2 | 162 | 81 | ||
| GenBank mRNA | JU336705.1 | - | 176 | 81 | ||
| JU470603.1 | - | 150 | 81 | |||
| JV045808.1 | - | 148 | 81 | |||
More Information
Set Membership: |
|

UQCC2
Genomic Map