Assay Details
Gene Symbol: KCTD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266955.1 | NP_001253884.1 | 638 | 85 | ||
XM_028838414.1 | XP_028694247.1 | 230 | 85 | |||
XM_028838415.1 | XP_028694248.1 | 645 | 85 | |||
XM_028838416.1 | XP_028694249.1 | 1747 | 85 | |||
XM_028838417.1 | XP_028694250.1 | 1750 | 85 | |||
XM_028838418.1 | XP_028694251.1 | 765 | 85 | |||
XM_028838419.1 | XP_028694252.1 | 762 | 85 | |||
XM_028838421.1 | XP_028694254.1 | 1864 | 85 | |||
GenBank mRNA | JU320215.1 | - | 638 | 85 | ||
JV668002.1 | - | 560 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028838414.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
