Assay Details
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260915.1 | NP_001247844.1 | 728 | 133 | ||
GenBank mRNA | JU337259.1 | - | 433 | 133 | ||
JU477966.1 | - | 408 | 133 | |||
JV048806.1 | - | 728 | 133 | |||
JV634365.1 | - | 748 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
