Assay Details
| Gene Symbol: SEC61A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266491.3 | NP_001253420.1 | 559 | 125 | ||
| GenBank mRNA | JU335252.1 | - | 584 | 125 | ||
| JU472613.1 | - | 545 | 125 | |||
| JV047518.1 | - | 560 | 125 | |||
| JV549610.1 | - | 612 | 125 | |||
More Information
Set Membership: |
|

SEC61A2
Genomic Map