Assay Details
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091148.4 | XP_001091148.3 | 1464 | 145 | ||
XM_015126826.2 | XP_014982312.1 | 1442 | 145 | |||
GenBank mRNA | JU325623.1 | - | 1154 | 145 | ||
JV668364.1 | - | 1416 | 145 |
Gene Symbol: FANCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU325622.1 | - | 1154 | 145 | ||
JU474667.1 | - | 5200 | 145 | |||
JU474668.1 | - | 5272 | 145 | |||
JU474670.1 | - | 5200 | 145 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
