Assay Details
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356559.2 | NP_001343488.1 | 1514 | 83 | ||
XM_028831572.1 | XP_028687405.1 | 1508 | 83 | |||
XM_028831573.1 | XP_028687406.1 | 1508 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
