Assay Details
Gene Symbol: PPP2R5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266743.2 | NP_001253672.1 | 421 | 74 | ||
XM_015143841.2 | XP_014999327.1 | 972 | 74 | |||
XM_015143842.2 | XP_014999328.1 | 975 | 74 | |||
XM_015143845.2 | XP_014999331.1 | 681 | 74 | |||
XM_015143846.2 | XP_014999332.1 | 343 | 74 | |||
GenBank mRNA | JU335302.1 | - | 455 | 74 | ||
JU473116.1 | - | 752 | 74 | |||
JV043507.1 | - | 421 | 74 | |||
JV718149.1 | - | 420 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
