Assay Details
| Gene Symbol: CYB5R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193767.2 | NP_001180696.1 | 1241 | 63 | ||
| XM_015136766.2 | XP_014992252.1 | 1331 | 63 | |||
| XM_015136768.2 | XP_014992254.1 | 1164 | 63 | |||
| GenBank mRNA | JU470893.1 | - | 1233 | 63 | ||
| JV703881.1 | - | 1228 | 63 | |||
More Information
Set Membership: |
|

CYB5R4
Genomic Map