Assay Details
| Gene Symbol: SFTPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042759.1 | NP_001036224.1 | 611 | 110 | ||
| XM_028852250.1 | XP_028708083.1 | 523 | 110 | |||
| GenBank mRNA | U06694.1 | - | 611 | 110 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SFTPC
Genomic Map